Masterflex® Sanitary Fitting End Caps, Polysulfone, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Close off your sanitary fluid line while creating a view port.
- Create a viewing port into your sanitary application with transparent end caps
- Constructed of polysulfone for enhanced strength and durability
- Fully compatible with other Tri-Clamp® connections
These end caps play an important role in the sanitation process, allowing an open end to be sanitary sealed preventing additional debris from accumulating while creating a view port for sample viewing. Choose from a wide range of end cap sizes, ranging from 1½” to 8”.
- Environmentally Preferable:
Specifications
- Max. temperature:
- 149 °C (300 °F)
- 149 °C (300 °F)
- 149 °C (300 °F)
- 149 °C (300 °F)
- 149 °C (300 °F)
- 149 °C (300 °F)
- Min. temperature:
- –100 °C (–150 °F)
- –100 °C (–150 °F)
- –100 °C (–150 °F)
- –100 °C (–150 °F)
- –100 °C (–150 °F)
- –100 °C (–150 °F)
- Sterilisation:
- Gamma irradiation and autoclave
- Gamma irradiation and autoclave
- Gamma irradiation and autoclave
- Gamma irradiation and autoclave
- Gamma irradiation and autoclave
- Gamma irradiation and autoclave
- Warranty:
- Manufacturer defect