Order Entry
India
ContactUsLinkComponent
Parker Hannifin Union Tee Compression Fittings
Parker Hannifin Union Tee Compression Fitting, Stainless Steel, 1/4" OD; Ea
Catalog # MFLX30486-27
undefined
Parker Hannifin Union Tee Compression Fitting, Stainless Steel, 1/4" OD; Ea
Catalog # MFLX30486-27
Specifications
  • Connection Type:
    Compression Union
  • Flow pattern:
    T
  • Description:
    Union Tee Compression Fitting, Stainless Steel, ¼" OD; Ea
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
    3 Months
  • Pk:
    1
  • Compression size / tube ext.Ø (port 3):
    ¼"
  • Fitting material:
    Stainless Steel 316
Specifications
About this item
Get a secure seal at higher pressures.
  • Quick and easy assembly without tools
Compression fittings offer a more permanent installation. Choose the size and material to suit your application needs.
  • Connection Type:
    Compression Union
  • Flow pattern:
    T
  • Description:
    Union Tee Compression Fitting, Stainless Steel, ¼" OD; Ea
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
    3 Months
  • Pk:
    1
  • Compression size / tube ext.Ø (port 3):
    ¼"
  • Fitting material:
    Stainless Steel 316