Order Entry
India
ContactUsLinkComponent
Antistatic Grounding Set, 3 Color Coded Copper Cables with Clamps
Burkle 5602-1000 Antistatic grounding set, 3 color coded copper cables with clamps
Catalog # MFLX06043-90
15 month warranty
undefined
Burkle 5602-1000 Antistatic grounding set, 3 color coded copper cables with clamps
Catalog # MFLX06043-90
15 month warranty
Specifications
  • Description:
    Antistatic grounding set, 3 color coded copper cables with clamps
  • Warranty:
    15 Months
  • Pk:
    1
Specifications
About this item
Ensures easy and reliable earthing.
  • Keep employees safe from fire or an explosion
When withdrawing flammable liquids, the container and pump must be grounded. Use the Burkle antistatic set to ground the pump to prevent possible electrostatic charges.
Delivery: Includes three colour-coded connecting copper cables with clamps in a handy plastic case.
  • Description:
    Antistatic grounding set, 3 color coded copper cables with clamps
  • Environmentally Preferable:
  • Warranty:
    15 Months
  • Pk:
    1