Order Entry
India
ContactUsLinkComponent
LinkTech 40CB Series Quick-Disconnect Fittings Body, Hose Barb with Valve
undefined
LinkTech 40CB Series Quick-Disconnect Fittings Body, Hose Barb with Valve
  • Fitting depth:
  • 23 mm (0,90625")
  • 23 mm (0,90625")
  • Max. operating pressure:
  • 250 psi
  • 250 psi
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • O-Ring material:
  • Buna-N
  • Buna-N
  • Valve material:
  • Acetal (POM)
  • Acetal (POM)
  • Valve spring material:
  • 316 SS
  • 316 SS
  • Warranty:
  • 1 year
  • 1 year
Frequently Bought Together