CELLTREAT Scientific Products Micro and Macro Centrifuge Tubes
Supplier: Avantor Fluid Handling
Certificates
About this item
LFlat-top cap offers easy one-handed opening and closing.
- Temperature range of -80 to 120 °C
- Feature a frosted marking spot, engraved graduations, and an attached polypropylene flat-top cap
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX06336-23
- MFLX06336-17
- MFLX06336-24
- Special feature:
- Nonsterile; skirted bottom
- Nonsterile; puncture membrane top
- Sterile; puncture membrane top