Masterflex® Sterile Vacuum Filtration Systems, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Ideal for sterile filtration of cell culture media.
- Polyethersulfone (PES) membrane provides fast flow rates and low binding
- Features a 0.22 μm PES membrane pore
- Sterilized by gamma irradiation and packaged in individual bags
- Environmentally Preferable:
Frequently Bought Together