Order Entry
India
ContactUsLinkComponent
ASCO Series 212 Water Conditioning and Purification Valves
undefined
ASCO Series 212 Water Conditioning and Purification Valves
  • Approval:
  • NSF
  • NSF
  • NSF
  • NSF
  • Max. pressure:
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • Max. temperature:
  • 50 °C (122 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Media compatibility:
  • Water based liquids
  • Water based liquids
  • Water based liquids
  • Water based liquids
  • Orifice:
  • 25,4 mm (1")
  • 19,05 mm (0,75")
  • 19,05 mm (0,75")
  • 19,05 mm (0,75")
  • Power:
  • 120 VDC
  • 120 VDC
  • 120 VDC
  • 120 VDC
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together