Order Entry
India
ContactUsLinkComponent
Masterflex® Personal Microcentrifuge Replacement Rotors, Avantor®
Masterflex® Personal Microcentrifuge Replacement Rotors, Avantor®
  • Model:
  • 17310-40
  • 17310-45
  • No. of tubes accepted:
  • 6
  • 2
  • Tube sizes accepted:
  • 1,5 or 2,0 ml microtubes
  • Strip tube
  • Rotor type:
  • Fixed-angle
  • Fixed-angle
  • Min. RPM:
  • 0 rpm
  • 0 rpm
  • Max. RPM:
  • 6000 rpm
  • 6000 rpm
  • Warranty:
  • 1 year
  • 1 year
Frequently Bought Together