Eppendorf 5810 Series Multi-Purpose Centrifuge Rotors
Change your centrifuge's capapcity with these rotors.
- Environmentally Preferable:
- Catalog No:
- MFLX02570-67
- MFLX02570-69
- No. of tubes accepted:
- 4
- 4
- Tube sizes accepted:
- 500 ml
- 250 ml
- Rotor type:
- Swing-out
- Swing-out
- Warranty:
- 1 year
- 1 year
Frequently Bought Together