Order Entry
India
ContactUsLinkComponent
CPC® SnapQuik® Quick-Disconnect Fittings
CPC® SnapQuik® Quick-Disconnect Fittings
Certificates
undefined
CPC® SnapQuik® Quick-Disconnect Fittings
Specifications
  • Max. operating pressure:
  • 15 psi
  • 15 psi
  • 15 psi
  • 15 psi
  • Max. temperature:
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • Min. temperature:
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • Seal material:
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year