Eppendorf 5424 Variable-Speed Refrigerated Microcentrifuges
Product Details & Documents
Added versatility to spin microcentrifuge tubes, miniprep spin columns, and PCR tube strips up to 15,000 rpm.
- Lid automatically opens at end of run for easy sample removal
- Suitable for use in cold rooms
- Whisper-quiet operation
Unit features microprocessor-based control with five-digit speed display of time and speed/g force. Easy-to-access control options include short-spin speed setting, soft-braking, “At Set” timer function, and adjustable volume alarm. Smooth keypad provides easy wipe-down cleaning. Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise. Order optional rotors, below right, for added versatility.
Delivery information: Includes 24 x 1.5/2.0 ml fixed-angle rotor with aeresol-tight lid and power cord with plug.
Specifications
- Description:Refrigerated Microcentrifuge; 24 x 2 mL Rotor, 230 VAC
- Rotors included:Yes
- Tube capacity:24 x 1,5/2,0 mL
- Warranty:2 years
- Max. speed:15000 rpm
- Min. temperature:–10 °C (14 °F)
- Max. RCF:21130 ×g
- Noise level:<54 dB(A)
- Rotor type:Fixed-Angle
- Cat. No.:MFLX02580-06
- Max. capacity:48 ml
- Length:26 cm (10")
- Max. temperature:40 °C (104 °F)
- Height:48 cm (18,9062")
- Speed control:Touch Pad
- Temperature control:Digital
- Width:29 cm (11,4062")
- Power:230 VAC, 50/60 Hz
Accessories
Product Information
- Catalog #AvailabilityPrice
- Adapter for fixed-angle and aerosol rotors; 0,2 PCR, 6/packAdapter for fixed-angle and aerosol rotors; 0,2 PCR, 6/packMFLX02570-58Log in for availability1 * 6 items
Product Information
- Catalog #AvailabilityPrice
- Adapter for fixed-angle and aerosol rotors; 0,5 mL tubes and BD Microtainer tubesAdapter for fixed-angle and aerosol rotors; 0,5 mL tubes and BD Microtainer tubesMFLX02580-57Log in for availability1 * 1 items
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



