Order Entry
India
ContactUsLinkComponent
Masterflex® Piston Pumps, Fixed Speed, for Series Q Pump Heads, Avantor®
undefined
Masterflex® Piston Pumps, Fixed Speed, for Series Q Pump Heads, Avantor®
  • Accuracy:
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Min. input mA:
  • 4 mA
  • 4 mA
  • 4 mA
  • 4 mA
  • 4 mA
  • 4 mA
  • Max. input mA:
  • 20 mA
  • 20 mA
  • 20 mA
  • 20 mA
  • 20 mA
  • 20 mA
  • Motor type:
  • AC Shaded Pole, IP 22
  • AC Shaded Pole, IP 22
  • AC Shaded Pole, IP 22
  • AC Shaded Pole, IP 22
  • AC Shaded Pole, IP 22
  • AC Shaded Pole, IP 22
  • Motor hp:
  • 1/20
  • 1/20
  • 1/20
  • 1/20
  • 1/20
  • 1/20
  • Power cord:
  • European
  • European
  • North American
  • North American
  • North American
  • North American
Frequently Bought Together