Order Entry
India
ContactUsLinkComponent
Antimicrobial Pleated Polyester Process Cartridges
Antimicrobial Pleated Polyester Filter Cartridge; 9.625" L, 50 um (nominal)
Catalog # MFLX29808-42
undefined
Antimicrobial Pleated Polyester Filter Cartridge; 9.625" L, 50 um (nominal)
Catalog # MFLX29808-42

Specifications

  • Filter media:
    Polyester
  • Micron Rating:
    50
  • Description:
    Antimicrobial Pleated Polyester Filter Cartridge; 9,625" L, 50 um (nominal)
  • Warranty:
    1 year
  • Pk:
    1
  • Filter length:
    9,625"

Specifications

About this item

Antimicrobial filter ensures long-lasting protection against algae, odor-causing bacteria, mold, mildew, and biofilms.

  • Constructed of pleated polyester with interwoven silver zeolite fibers

Antimicrobial Cartridges are pleated polyester with interwoven silver zeolite fibers. The interwoven fibers ensure long-lasting protection against algae, odor-causing bacteria, mold, mildew, and biofilms that might otherwise grow on the cartridge surface.

  • Filter media:
    Polyester
  • Micron Rating:
    50
  • Description:
    Antimicrobial Pleated Polyester Filter Cartridge; 9,625" L, 50 um (nominal)
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
  • Filter length:
    9,625"