Order Entry
India
ContactUsLinkComponent
Parker Hannifin TrueSeal PVDF Push-to-Connect Fittings, Male Threaded Adapters
Parker Hannifin TrueSeal PVDF Push-to-Connect Fittings, Male Threaded Adapters
Certificates
undefined
Parker Hannifin TrueSeal PVDF Push-to-Connect Fittings, Male Threaded Adapters
Specifications
  • Max. operating pressure:
  • 300 psi
  • 300 psi
  • 300 psi
  • Max. temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Max. vacuum:
  • 762 mm Hg (30" Hg)
  • 762 mm Hg (30" Hg)
  • 762 mm Hg (30" Hg)
  • Min. temperature:
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • Seal material:
  • FKM
  • FKM
  • FKM
  • Sterilisation:
  • Not recommended
  • Not recommended
  • Not recommended
  • Warranty:
  • 1 year
  • 1 year
  • 1 year