Order Entry
India
ContactUsLinkComponent
Bulkhead Fitting, PVDF, 1" NPT(F) Port X 2-5/8" OD
Catalog # MFLX34031-23
undefined
Bulkhead Fitting, PVDF, 1" NPT(F) Port X 2-5/8" OD
Catalog # MFLX34031-23
Specifications
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    1"
  • Description:
    Bulkhead Fitting, PVDF, 1" NPT(F) Port X 2-⅝" OD
  • Bulkhead outer diameter:
    2,625
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    2 years
  • Pk:
    1
  • Fitting material:
    PVDF
Specifications
About this item
Constructed of high purity PVDF ideal for chemically aggressive applications.
  • Constructed of durable PVDF
These bulkhead fittings allow quick, easy connection of our Schedule 80 PTFE threaded pipe system, or any NPT pipe system to tanks of any material: plastic, metal, or lined. The fittings can be installed without adapters in either flat wall tanks, or many cylindrical tanks.
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    1"
  • Description:
    Bulkhead Fitting, PVDF, 1" NPT(F) Port X 2-⅝" OD
  • Environmentally Preferable:
  • Bulkhead outer diameter:
    2,625
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    2 years
  • Pk:
    1
  • Fitting material:
    PVDF