Order Entry
India
ContactUsLinkComponent
Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Certificates
undefined
Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Specifications
  • Inlet connection:
  • 1" Tri-Clamp®
  • 1" Tri-Clamp®
  • 1" Tri-Clamp®
  • 1" Tri-Clamp®
  • Max. temperature:
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • Max. viscosity:
  • 40000 cP
  • 40000 cP
  • 40000 cP
  • 40000 cP
  • Min. flow rate:
  • 0,08 L/min (0,021 gal./min)
  • 0,011 L/min (0,003 gal./min)
  • 0,045 L/min (0,012 gal./min)
  • 0,003 L/min (0,0008 gal./min)
  • Motor type:
  • Variable-frequency drive
  • 56C
  • Variable-frequency drive
  • 56C
  • Motor hp:
  • 1/2
  • 1/2
  • 1/2
  • Outlet connection:
  • 3/4" Tri-Clamp®
  • 3/4" Tri-Clamp®
  • 3/4" Tri-Clamp®
  • 3/4" Tri-Clamp®
  • Suction lift, wet:
  • 2700 mm of H₂O (108" H₂O); 559 mm Hg (7,9" Hg)
  • 2700 mm of H₂O (108" H₂O); 559 mm Hg (7,9" Hg)
  • 2700 mm of H₂O (108" H₂O); 559 mm Hg (7,9" Hg)
  • 2700 mm of H₂O (108" H₂O); 559 mm Hg (7,9" Hg)
  • Wetted materials:
  • 316 Stainless Stele, Therban®
  • 316 Stainless Stele, Therban®
  • 316 Stainless Stele, Therban®
  • 316 Stainless Stele, Therban®
  • Warranty:
  • 3 years
  • 3 years
  • 3 years
  • 3 years