Animal-Derived Ingredient Free Sanitary Adapters
Supplier: Avantor Fluid Handling
Certificates
About this item
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow, no particulate build-up and easy cleaning.
Made from animal-derived ingredient-free polypropylene and PVDF, these adapters may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
- Environmentally Preferable:
Specifications
- Sterilisation:
- Gamma only
- Gamma only
- Gamma only
- Gamma only
- Gamma only