Order Entry
India
ContactUsLinkComponent
CPC® Miniature Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
CPC® Miniature Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Certificates
undefined
CPC® Miniature Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Specifications
  • Max. operating pressure:
  • 45 psi
  • 45 psi
  • Max. temperature:
  • 49 °C (120 °F)
  • 49 °C (120 °F)
  • 49 °C (120 °F)
  • Min. temperature:
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • Seal material:
  • EPDM
  • EPDM
  • EPDM
  • Valve spring material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year