Masterflex® Female luer x 1/4-28 UNF bottom sealing thread, PP, 25/pk 45503-76
Catalog # MFLX45503-76
Supplier: Avantor Fluid Handling
CE certificate
Avantor exclusive
Specifications
- Connection Type:Female Luer to Thread
- Flow pattern:Straight
- Description:Female luer × ¼-28 UNF bottom sealing thread, PP, 25/pk 45503-76
- Warranty:3 Months
- Pk:25
- Fitting material:Polypropylene
Specifications
About this item
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- Connection Type:Female Luer to Thread
- Flow pattern:Straight
- Description:Female luer × ¼-28 UNF bottom sealing thread, PP, 25/pk 45503-76
- Environmentally Preferable:
- Warranty:3 Months
- Pk:25
- Fitting material:Polypropylene