Masterflex® Female luer x female luer adapter, PP, 25/pk 45500-22
MFLX45500-22
CE certificate
Avantor exclusive
- Connection Type:Female Luer Union
- Flow pattern:Straight
- Description:Female luer x female luer adapter, PP, 25/pk 45500-22
- Warranty:3 Months
- Pk:25
- Fitting material:Polypropylene
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.
- Connection Type:Female Luer Union
- Flow pattern:Straight
- Description:Female luer x female luer adapter, PP, 25/pk 45500-22
- Environmentally Preferable:
- Warranty:3 Months
- Pk:25
- Fitting material:Polypropylene