Order Entry
India
ContactUsLinkComponent
Masterflex® Adapter Fittings, Female NPT Threaded to Bulkhead, Straight, Avantor®
undefined
Masterflex® Adapter Fittings, Female NPT Threaded to Bulkhead, Straight, Avantor®
  • Model:
  • 06349-10
  • 06349-12
  • 06349-11
  • Bulkhead outer diameter:
  • 0,625
  • 0,875
  • 0,75
  • Fitting depth:
  • 7,9375 mm
  • 6,35 mm
  • 11,1125 mm
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together