Replacement PFA Compression Ferrule Nut and Assemblies
Supplier: Avantor Fluid Handling
About this item
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
2 Options Available Below
Product Details & Documents
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX31320-46
- MFLX31321-89
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Warranty:
- 18 months
- 18 months
Specifications
Product Family Options
Product Information
- Connection TypeFitting materialFlow patternAvailabilityPrice
- MFLX31320-46Ferrule plug, PFA, ¼"Specifications:CE certificate
More
Specifications
Cat. No.MFLX31320-46SterilisationAutoclave or ethylene oxide (ETO)Warranty18 monthsCE certificate - MFLX31321-89Compression Nut Assembly, PFA, ¾"Compression Nut Assembly, PFA, ¾"Compression Nut Assembly with FerrulePFAStraightLog in for availability1 * 1 itemsRs.6,800.00Specifications:
More
Specifications
Cat. No.MFLX31321-89SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months