Order Entry
India
ContactUsLinkComponent
Two-Way, Three-Way, and Sample Diaphragm Valves
undefined
Two-Way, Three-Way, and Sample Diaphragm Valves
  • Max. pressure:
  • 60 psi
  • 100 psi
  • 100 psi
  • 45 psi
  • Max. temperature:
  • 107 °C (225 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 93 °C (200 °F)
  • Orifice:
  • 9,53 mm (0,375")
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • Wetted materials:
  • PFA
  • Warranty:
  • 1 year
  • 18 months
  • 18 months
  • 1 year
  • 18 months
  • 18 months
Frequently Bought Together