Order Entry
India
ContactUsLinkComponent
Parker Hannifin® Flare Union Elbows, PFA
Parker Hannifin Flare Union Fitting, Elbow, 3/8", PFA; 1/Ea
Catalog # MFLX30490-48
undefined
Parker Hannifin Flare Union Fitting, Elbow, 3/8", PFA; 1/Ea
Catalog # MFLX30490-48
Specifications
  • Connection Type:
    Flare Union
  • Flow pattern:
    Elbow
  • Description:
    Flare union Elbow, ⅜", PFA, Each
  • Warranty:
    3 Months
  • Pk:
    1
  • Fitting material:
    PFA
Specifications
About this item
Leak-tight and high-pressure connection for corrosive chemical applications.
  • Constructed of durable PFA
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.
  • Connection Type:
    Flare Union
  • Flow pattern:
    Elbow
  • Description:
    Flare union Elbow, ⅜", PFA, Each
  • Environmentally Preferable:
  • Warranty:
    3 Months
  • Pk:
    1
  • Fitting material:
    PFA