Order Entry
India
ContactUsLinkComponent
Finish Thompson Economical Drum Pump Tube Sets
Finish Thompson Economical Drum Pump Tube Sets
Certificates
undefined
Finish Thompson Economical Drum Pump Tube Sets
Specifications
  • Max. temperature:
  • 66 °C (150 °F)
  • 100 °C (212 °F)
  • 71 °C (160 °F)
  • Outlet connection:
  • 3/4" hose barb
  • 3/4" hose barb
  • 3/4" hose barb
  • Tube ext.Ø, inlet:
  • 3,175 cm (1,25")
  • 3,175 cm (1,25")
  • 3,175 cm (1,25")
  • Tube length, inlet:
  • 69 cm (27")
  • 102 cm (40")
  • 102 cm (40")
  • Wetted materials:
  • PP tube, 316 SS shaft, FKM, and PTFE internals
  • 316 SS tube, 316 SS shaft, FKM, PTFE, ETFE internals
  • Pure PP/PVDF tube, Alloy 625 shaft, FKM, PTFE, ETFE internals
  • Warranty:
  • 1 year
  • 1 year
  • 1 year