Order Entry
India
ContactUsLinkComponent
Parker Hannifin® Metal Needle Valves
undefined
Parker Hannifin® Metal Needle Valves
  • Max. pressure:
  • 5000 psi
  • 3000 psi
  • 5000 psi
  • Max. temperature:
  • 232 °C (450 °F)
  • 232 °C (450 °F)
  • 232 °C (450 °F)
  • Orifice:
  • 4,47 mm (0,176")
  • 5,7912 mm (0,228")
  • 7,9248 mm (0,312")
  • Tubing ext.Ø:
  • 0,25"
  • 0,375"
  • 0,5"
  • Wetted materials:
  • Stainless Steel
  • Brass
  • Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together