Cast iron float and thermostatic steam traps (in-line horizontal configuration)
Supplier: Avantor Fluid Handling
Certificates
About this item
Keep steam space free of condensate to provide maximum system efficiency.
- Constructed of durable cast iron
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
Air heating coils
Heat exchangers
Steam main drip stations
Small process equipment, particularly when controlled by a modulating temperature control valve
Replacement of less efficient inverted bucket traps on heating equipment
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX07600-52
- MFLX07600-50
- MFLX07600-46
- Max. operating pressure:
- 125 psi
- 75 psi
- 15 psi
- Max. temperature:
- 232 °C of superheat (450 °F of superheat)
- 232 °C of superheat (450 °F of superheat)
- 232 °C of superheat (450 °F of superheat)
- Warranty:
- 1 year
- 1 year
- 1 year