Masterflex® Ismatec® ISM734B Tubing Bed Pump Head Adapter Plate for Tubing Bed Pump Heads
Catalog # MFLX78002-80
Supplier: Avantor Fluid Handling
Specifications
- Description:Tubing Bed Pump Head Adapter Plate for Tubing Bed Pump Heads
- Pk:1
Specifications
About this item
Adapt your drive to accept Ismatec® tubing bed pump heads.
- Suitable for use with Ismatec® tubing bed pump heads
This is the mounting plate needed to attach an Ismatec® tubing bed pump head to an Ismatec® Drive. Suitable for use with the 07611-10 and -15 pump heads on the 78002-Series drives.
- Description:Tubing Bed Pump Head Adapter Plate for Tubing Bed Pump Heads
- Environmentally Preferable:
- Pk:1