Order Entry
India
ContactUsLinkComponent
Eppendorf 5804 and 5810 Series Multi-Purpose Centrifuge Rotors
Eppendorf 5804 and 5810 Series Multi-Purpose Centrifuge Rotors
  • No. of tubes accepted:
  • 6
  • 4
  • Tube sizes accepted:
  • 100 ml
  • 100 ml
  • Rotor type:
  • Fixed-angle
  • Swing-out
  • Max. RCF:
  • 15500 ×g
  • 4500 ×g
  • Warranty:
  • 2 years
  • 2 years
Frequently Bought Together