Order Entry
India
ContactUsLinkComponent
Medium, Ham's F-12, Corning®
Medium, Ham's F-12, Corning®
Catalog # 50-040-PB
Supplier:  Corning
CAS Number:  
undefined
Medium, Ham's F-12, Corning®
Catalog # 50-040-PB
Supplier:  Corning
CAS Number:  

Specifications

  • Media Format:
    Powder
  • Pk:
    10 L
  • Description:
    Ham's F-12 medium, powder with L-glutamine without sodium bicarbonate
  • Storage temperature:
    Refrigerate

Specifications

About this item

Ham's F-12 media was originally formulated for the serum-free culture of CHO cells, and is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.

  • Low endotoxin - < 0,25 EU/ml
  • Mycoplasma tested
  • Liquid media formulations are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

This medium is also suitable for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos.