Order Entry
India
ContactUsLinkComponent
Masterflex® High-Efficiency Absolute-Rated Filter Bags, Polypropylene, Avantor®
undefined
Masterflex® High-Efficiency Absolute-Rated Filter Bags, Polypropylene, Avantor®
  • Filter diameter:
  • 7"
  • 7"
  • 7"
  • Filtration area:
  • 0,409 m² (4,4 ft.², 633,6 in²)
  • 0,409 m² (4,4 ft.², 633,6 in²)
  • 0,409 m² (4,4 ft.², 633,6 in²)
  • Max. temperature:
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • Pore size:
  • 1 µm
  • 25 µm
  • 50 µm
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together