Dissolved Oxygen Control Gas Diffuser Aeration System
About this item
Increase oxygen into your water.
- Ideal for water and wastewater treatment
- Provides oxygenation and mixing capabilities essential for biological treatment processes
Diffuser aeration systems provide increased oxygen content to wastewater in treatment facilities. This process creates an environment where microorganisms biologically decompose soluble organic contaminants in wastewater.
Diffusers are also used to diffuse digester gases such as hydrogen sulfide and sulphur dioxide; for the introduction of ozone for the purpose of oxidizing and destroying bacterial and organic chemical contaminants; and for carbon dioxide injection for recarbonization and pH adjustment. Order air blowers separately to complete your system.
10 Options Available Below
Product Details & Documents
Increase oxygen into your water.
- Ideal for water and wastewater treatment
- Provides oxygenation and mixing capabilities essential for biological treatment processes
Diffuser aeration systems provide increased oxygen content to wastewater in treatment facilities. This process creates an environment where microorganisms biologically decompose soluble organic contaminants in wastewater.
Diffusers are also used to diffuse digester gases such as hydrogen sulfide and sulphur dioxide; for the introduction of ozone for the purpose of oxidizing and destroying bacterial and organic chemical contaminants; and for carbon dioxide injection for recarbonization and pH adjustment. Order air blowers separately to complete your system.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX70025-00
- MFLX70025-02
- MFLX70025-06
- MFLX70025-10
- MFLX70025-12
- MFLX70025-20
- MFLX70025-22
- MFLX70025-24
- MFLX70025-28
- MFLX70025-30
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifications
Product Family Options
Product Information
- MaterialThread size NPTMin. flow rangeMax. flow rangeAvailabilityPrice
- MFLX70025-00Diffuser, coarseSpecifications:
More
Specifications
Cat. No.MFLX70025-00Warranty1 year - MFLX70025-02Diffuser, coarseDiffuser, coarseStainless steel 3040,375"0,5 CFM3 CFMLog in for availability1 * 1 itemsRs.12,100.00Specifications:
More
Specifications
Cat. No.MFLX70025-02Warranty1 year - MFLX70025-06Diffuser, coarseSpecifications:
More
Specifications
Cat. No.MFLX70025-06Warranty1 year - MFLX70025-10Diffuser, coarseSpecifications:
More
Specifications
Cat. No.MFLX70025-10Warranty1 year - MFLX70025-12Diffuser, coarseSpecifications:
More
Specifications
Cat. No.MFLX70025-12Warranty1 year - MFLX70025-20Diffuser, fineSpecifications:
More
Specifications
Cat. No.MFLX70025-20Warranty1 year - MFLX70025-22Diffuser, fineSpecifications:
More
Specifications
Cat. No.MFLX70025-22Warranty1 year - MFLX70025-24DiffuserSpecifications:
More
Specifications
Cat. No.MFLX70025-24Warranty1 year - MFLX70025-28Diffuser low-flowSpecifications:
More
Specifications
Cat. No.MFLX70025-28Warranty1 year - MFLX70025-30Diffuser check valve, fineSpecifications:
More
Specifications
Cat. No.MFLX70025-30Warranty1 year