Order Entry
India
ContactUsLinkComponent
Masterflex® Stopcocks for Glass and Rigid Tubing, Avantor®
Masterflex® Two-Way Stopcock for Glass and Rigid Tubing, 10 mm tube OD
  MFLX30503-04
undefined
Masterflex® Two-Way Stopcock for Glass and Rigid Tubing, 10 mm tube OD
  MFLX30503-04
  • Connection type:
    Compression
  • Description:
    Two-Way Stopcock for Glass and Rigid Tubing, 10 mm tube OD
  • Pk:
    1
Easily connect to rigid tubing.
Stopcocks have wetted parts constructed of PFA. An O-ring made of Viton® protects the glass tube when you tighten the compression nut. Stopcocks also accept rigid tubing such as PTFE.
Available in two-way and three-way styles.
  • Connection type:
    Compression
  • Description:
    Two-Way Stopcock for Glass and Rigid Tubing, 10 mm tube OD
  • Environmentally Preferable:
  • Pk:
    1