Order Entry
India
ContactUsLinkComponent
Eldex High-Pressure Piston Pumps
Eldex High-Pressure Piston Pumps
Certificates
undefined
Eldex High-Pressure Piston Pumps
Specifications
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 2500 psi
  • 1000 psi
  • 5000 psi
  • Max. temperature:
  • 120 °C (248 °F)
  • 120 °C (248 °F)
  • 120 °C (248 °F)
  • Power cord:
  • 7-ft, three-wire cord with plug
  • 7-ft, three-wire cord with plug
  • 7-ft, three-wire cord with plug
  • Repeatability:
  • ±0,3%
  • ±0,3%
  • ±0,3%
  • Wetted materials:
  • 316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
  • 316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
  • 316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
  • Warranty:
  • 1 year
  • 1 year
  • 1 year