Order Entry
India
ContactUsLinkComponent
Masterflex® I/P® Variable-Speed Hazardous-Duty Drives, Avantor®
Masterflex® I/P® Variable-Speed Hazardous-Duty Drive, 10 to 430 rpm; 115V
  MFLX07583-50
2 year warranty
ISO9001
undefined
Masterflex® I/P® Variable-Speed Hazardous-Duty Drive, 10 to 430 rpm; 115V
  MFLX07583-50
2 year warranty
ISO9001

 

  • Maximum Flow Rate:
    11500 ml/min
  • Power:
    115 VAC; 60 Hz; 4,7 A
  • Control type:
    Analog Variable Speed
  • Pump series:
    I/P
  • Description:
    I/P variable-speed hazardous-duty drive, 10-430 rpm, 115V.
  • Drive height:
    223 mm (8,75")
  • Drive IP rating:
    IP 21
  • Drive length:
    560 mm (22,0625")
  • Drive width:
    346 mm (13,625")
  • Min. RPM:
    10 rpm
  • Motor hp:
    1/3
  • Open-head sensor:
    No
  • Reversible motor:
    No
  • Speed control:
    ±10%
  • Type of pump heads accepted:
    I/P: Standard, Easy-Load, High-Performance; L/S: Standard, Quick Load, Easy-Load, Easy-Load II, High-Performance, Cartridge
  • Motor:
    190 W
  • Speed:
    Variable
  • Warranty:
    2 years
  • Pk:
    1
  • Housing material:
    Painted steel
  • Max. RPM:
    430 rpm
  • No. of pump heads accepted:
    2

 

 

Pump safely in hazardous environments.

  • Use either L/S or I/P pump head on drive
  • L/S flow rate 0.6 to 2500 ml/min or I/P flow rate 60 to 8800 ml/min— flow rate depends on drive rpm, pump head, and tubing size
  • Fully mechanical speed control—adjust with lever then lock setting
  • UL-listed for Class 1, Groups C and D, Division 1 hazardous locations

Choose one of these variable-speed hazardous-duty drives for hazardous locations where special electrical precautions are necessary. Power switch and line cord are not included. Safely connect power using your own protected connections.

  • Maximum Flow Rate:
    11500 ml/min
  • Power:
    115 VAC; 60 Hz; 4,7 A
  • Control type:
    Analog Variable Speed
  • Pump series:
    I/P
  • Description:
    I/P variable-speed hazardous-duty drive, 10-430 rpm, 115V.
  • Environmentally Preferable:
  • Drive height:
    223 mm (8,75")
  • Drive IP rating:
    IP 21
  • Drive length:
    560 mm (22,0625")
  • Drive width:
    346 mm (13,625")
  • Min. RPM:
    10 rpm
  • Motor hp:
    1/3
  • Open-head sensor:
    No
  • Reversible motor:
    No
  • Speed control:
    ±10%
  • Type of pump heads accepted:
    I/P: Standard, Easy-Load, High-Performance; L/S: Standard, Quick Load, Easy-Load, Easy-Load II, High-Performance, Cartridge
  • Motor:
    190 W
  • Speed:
    Variable
  • Warranty:
    2 years
  • Pk:
    1
  • Housing material:
    Painted steel
  • Max. RPM:
    430 rpm
  • No. of pump heads accepted:
    2