Order Entry
India
ContactUsLinkComponent
Masterflex® Quick-Disconnect Coupler Fittings, Hose Barbed, Straight, Avantor®
Masterflex® Fitting, LDPE, Straight, Hosebarb to Quick-Disconnect, 3/8" ID x 7/16" ID; 20/PK
Catalog # MFLX06290-15
undefined
Masterflex® Fitting, LDPE, Straight, Hosebarb to Quick-Disconnect, 3/8" ID x 7/16" ID; 20/PK
Catalog # MFLX06290-15
Specifications
  • Barb size/tubing int.Ø (port 1):
    ⅜"
  • Barb size/tubing int.Ø (port 2):
    ⁷⁄₁₆"
  • Connection Type:
    Quick-Disconnect to Barb
  • Flow pattern:
    Straight
  • Type:
    Open
  • Description:
    Barbed Straight Quick-Connect Couplers, 3/8 x ⁷⁄₁₆" ID, LDPE, 20/Pk
  • Seq:
    04
  • Warranty:
    1 year
  • Pk:
    20
  • Fitting material:
    LDPE
Specifications
About this item
Provides easy single-handed disconnection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Construction from low-density polyethylene (LDPE).
  • Barb size/tubing int.Ø (port 1):
    ⅜"
  • Barb size/tubing int.Ø (port 2):
    ⁷⁄₁₆"
  • Connection Type:
    Quick-Disconnect to Barb
  • Flow pattern:
    Straight
  • Type:
    Open
  • Description:
    Barbed Straight Quick-Connect Couplers, 3/8 x ⁷⁄₁₆" ID, LDPE, 20/Pk
  • Seq:
    04
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    20
  • Fitting material:
    LDPE