Order Entry
India
ContactUsLinkComponent
Masterflex®, Adapter Fittings, Compression to Male Threaded, Elbow, Avantor®
Masterflex®, Adapter Fittings, Compression to Male Threaded, Elbow, Avantor®
Certificates
undefined
Masterflex®, Adapter Fittings, Compression to Male Threaded, Elbow, Avantor®
Specifications
  • Model:
  • 30815-38
  • 30805-45
  • 30815-39
  • 30803-38
  • Orifice dimensions:
  • 8,73 mm (0,34375")
  • 11,9 mm (0,46875")
  • 9,53 mm (0,375")
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Warranty:
  • Out of box
  • Out of box
  • Out of box