Order Entry
India
ContactUsLinkComponent
Union Fitting, Male to Female Threaded Adapter, Elbow
Union Fitting, Male to Female Threaded Adapter, Elbow
Certificates
undefined
Union Fitting, Male to Female Threaded Adapter, Elbow
Specifications
  • Model:
  • 06546-14
  • 06546-16
  • 06349-41
  • 06349-43
  • 06349-42
  • 31320-90
  • 31320-88
  • 31320-89
  • 06546-13
  • Fitting depth:
  • 7,14 mm
  • 10,32 mm
  • 9,525 mm
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Warranty:
  • 1 year
  • 1 year
  • 1 year