Parker Hannifin British Standard Pipe Threaded (BSPP) Adapters
Supplier: Avantor Fluid Handling
Certificates
About this item
Convert NPT-threaded systems to BSPP threads—or visa versa—with these stainless steel fittings.
- Constructed of durable stainless steel
Use BSP parallel (BSPP) fittings for connections where pressure-tight joints are not made on the thread (as in connections with peripheral seals). Stainless steel fittings are chemically resistant.
- Environmentally Preferable:
Specifications
- Sterilisation:
- Ethylene oxide (EtO), autoclave, or gamma irradiation
- Ethylene oxide (EtO), autoclave, or gamma irradiation
- Ethylene oxide (EtO), autoclave, or gamma irradiation
- Ethylene oxide (EtO), autoclave, or gamma irradiation
- Thread size BSP(F) (port 1):
- 0,5"
- 0,25"
- 0,375"
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year