Pentair Dual-Layer Gradient-Density Filtration Catridges
Supplier: Avantor Fluid Handling
Certificates
About this item
Dual-layer design has a holding capacity of two to three times traditional cartridges.
- Constructed of polypropylene (PP)
Dual-layer cartridges are high-capacity cartridges that combine a prefiltration layer with a selective postfiltration layer. Each layer is a separate gradient-density filter, which reduces larger particles on the outer layers and finer particles on the inner layers. Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust and scale.
This design has a holding capacity that is two to three times traditional spun or wound designs. Catridges are made of bacteria and chemical-resistant polypropylene; suitable to 145 °F (63 °C). These cartridges are certified to NSF Standard 42 for materials.
- Environmentally Preferable:
Frequently Bought Together