Order Entry
India
ContactUsLinkComponent
Pentair Hardness/Iron Reduction Double-Open-End Liquid Filter Cartridges
Pentair Double open end hexametaphosphate crystal cartridge; 10"L
  MFLX29830-60
undefined
Pentair Double open end hexametaphosphate crystal cartridge; 10"L
  MFLX29830-60
  • Filter media:
    Hexametaphosphate
  • Flow rate:
    1,5 - 2,5 gal./min
  • Max. temperature:
    38 °C (100 °F)
  • Description:
    Double open end hexametaphosphate crystal cartridge; 10"L
  • Warranty:
    1 year
  • Pk:
    1
  • Filter length:
    10"
Prevents scale and rust buildup—ideal for water processing.
  • Constructed with Hexametaphosphate crystals
  • FDA-compliant
Hexametaphosphate Crystal Cartridge prevents scale and rust buildup — use with water heaters, air conditioning equipment, and many other types of water processing equipment. Food-grade hexametaphosphate dissolves slowly in water to treat scale, corrosion, and iron problems at various flow rates and feed concentrations. All materials are FDA-compliant.
Water being treated should have no more than 257 mg/L hardness, with a pH range of 6.5 to 9.0. Maximum temperature is 100 °F (38 °C). Cartridge is made from polypropylene (PP) shell with a polyester pad prefilter and a PP post filter.
  • Filter media:
    Hexametaphosphate
  • Flow rate:
    1,5 - 2,5 gal./min
  • Max. temperature:
    38 °C (100 °F)
  • Description:
    Double open end hexametaphosphate crystal cartridge; 10"L
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
  • Filter length:
    10"