Order Entry
India
ContactUsLinkComponent
Masterflex® Adapter Fittings, Compression to Male Threaded, Tee, Avantor®
Masterflex® Fitting, Polypropylene, Tee, Compression to Threaded Adapter, 3/8" OD x 3/8" OD x 1/4" NPT(M); 10/PK
Catalog # MFLX06388-40
undefined
Masterflex® Fitting, Polypropylene, Tee, Compression to Threaded Adapter, 3/8" OD x 3/8" OD x 1/4" NPT(M); 10/PK
Catalog # MFLX06388-40
Specifications
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    T
  • Description:
    Male compression run tees, PP, ⅜" OD x ¼" NPT(M), pack of 10
  • Seq:
    03
  • Model:
    06388-40
  • Orifice dimensions:
    0,34375"
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    Polypropylene
Specifications
About this item
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Choose nylon for good chemical resistance and higher temperature applications. Both nylon and polypropylene can be sterilized via ethylene oxide.
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    T
  • Description:
    Male compression run tees, PP, ⅜" OD x ¼" NPT(M), pack of 10
  • Seq:
    03
  • Environmentally Preferable:
  • Model:
    06388-40
  • Orifice dimensions:
    0,34375"
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    Polypropylene