Thermo Scientific Nalgene DS3147-0050 Conical Tube Adapter 6/pk
MFLX06327-79
- Description:Conical Tube AdapteR 6/pk
- Environmentally Preferable:
- Pk:6
- Warranty:1 year
Secure your Oak Ridge conical bottom tubes.
- Constructed of durable polycarbonate
- Max force of 50,000 x g
- Autoclavable
Easily remove the Thermo Scientific™ Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use. This autoclavable adapter must be used with Nalgene 35L conical Oak Ridge Tubes 06327-70 and -72 (tubes sold separately).
- Description:Conical Tube AdapteR 6/pk
- Environmentally Preferable:
- Pk:6
- Warranty:1 year