Accessory Heat sink for DC controller 70100-10
Catalog # MFLX70100-50
Supplier: Avantor Fluid Handling
Specifications
- Description:Accessory Heat sink for DC controller 70100-10
- Warranty:1 year
- Pk:1
Specifications
About this item
Opimize performance of your Variable-Speed Controller.
- Double maximum armature current and horsepower when drive is mounted on heat sink
- Use heat sink when the continuous armature current is above 5 ADC
Compatible with the Variable-Speed Controller model 70100-10, controller sold separately.
- Description:Accessory Heat sink for DC controller 70100-10
- Environmentally Preferable:
- Warranty:1 year
- Pk:1