Thermo Scientific Nalgene Snap-On Centrifuge Tube PP Closures
Pull tab makes it easy to remove.
- Autoclavable
These snap-on closures for plastic centrifuge tubes are easy to remove with a convenient pull tab. Designed to fit Nalgene round and conical centrifuge tubes. Not liquid tight at high speeds.
Thermo Scientific Nalgene™, Thermo Scientific Nunc International.
Warranty: 1 year
- Environmentally Preferable:
Frequently Bought Together