Order Entry
India
ContactUsLinkComponent
Thermo Scientific Nalgene Centrifuge Bottle Sealing Cap Assembly
Thermo Scientific Nalgene Centrifuge Bottle Sealing Cap Assembly, 38mm; 2/PK
  MFLX06327-57
Thermo Scientific Nalgene Centrifuge Bottle Sealing Cap Assembly, 38mm; 2/PK
  MFLX06327-57

 

  • Description:
    Centrifuge Bottle Sealing Cap Assembly, 38mm
  • Pk:
    2
  • For bottles:
    250 - 290 ml
  • Max. capacity:
    290 ml

 

 

Offers added level of reliability.

  • Autoclavable

Designed for Oak Ridge tubes and centrifuge bottles, the bottle sealing caps ensure vessels won’t leak at high speeds. Includes PP screw closure, TEFZEL® ETFE plugs and VITON® flouroelastomer O-rings.

Warranty: 1 year

  • Description:
    Centrifuge Bottle Sealing Cap Assembly, 38mm
  • Environmentally Preferable:
  • Pk:
    2
  • For bottles:
    250 - 290 ml
  • Max. capacity:
    290 ml