Pyrex Conical-Bottom Centrifuge Tubes, Glass
Supplier: Avantor Fluid Handling
Certificates
About this item
Tubes are manufactured from Pyrex® brand borosilicate glass for greater strength and clarity. Manufacturing process ensures that tubes are strengthened at stress points.
Note: To minimize the risks of tube breakage, use the proper adapters or cushions in centrifuge rotors.
- Environmentally Preferable:
Frequently Bought Together