Order Entry
India
ContactUsLinkComponent
10X Tris buffered saline, liquid, Corning®
10X Tris buffered saline, liquid, Corning®
  MDTC46-012-CM
 :  Corning
 :  
undefined
10X Tris buffered saline, liquid, Corning®
  MDTC46-012-CM
 :  Corning
 :  
  • Pk:
    6
  • Description:
    10X tris buffered saline, liquid, 1L
  • Environmentally Preferable:
  • Storage temperature:
    Ambient
  • Size:
    1 L
  • Shipping temperature:
    Ambient
Tris Buffered Saline (TBS) is an isotonic, non-toxic buffer used in some biomedical techniques to maintain pH within a relatively narrow range.
TBS can shift pH relatively dramatically as temperature changes, with the pKa declining approximately 0,03 units per degree Celsius rise in temperature.