Corning® Human recombinant albumin (from serum)
MDTC62-450-RF
: Corning
:
- Pk:1 g
- Protein/peptide type:Recombinant
- Source:Serum
- Species:Human
- Storage conditions:Refrigerate/Frozen
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
- Protein/peptide name:Albumin
- Shipping temperature:Ambient
Corning® cellgro® Albumin is a chemically-defined recombinant human serum albumin (rhHSA). Having an essential and non-essential fatty acid profile, this is a powerful media supplement that outperforms plasma-derived human serum albumin (pHSA), bovine serum albumin (BSA) and FBS. Some prominent applications for cellgro® rhAlbumin are as a supplement for CHO cell culture and for the culture of various other cell lines. This product is plant-derived and completely animal-origin free.