Lithium iodide, anhydrous ≥99.999% (trace metals basis)
Catalog # VWRMNP134525-100G
Supplier: VWR USA
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:99.999%
Specification Test Results
About this item
:
:133.85 g/mol
Melting Pt:446 °C
Density:3,49 g/mL
:ampouled under argon
EINECS:233-822-5
CAS Number:10377-51-2
Formula:ILi
MDL Number:MFCD00011092